Afu UDG, New England Biolabs
Supplier: New England Biolabs (NEB)
M0279S
101417-982EA 88 USD
101417-982
Afu UDG, New England Biolabs
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
A thermostable homolog of the E.coli Uracil-DNA Glycosylase (UDG) from Archaeoglobus fulgidus.
Afu UDG catalyzes the release of free uracil from uracil-containing DNA. Afu UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA.