T7 Express lysY Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)
C3010I
101417-910EA 208 USD
101417-910
T7 Express lysY Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
Enhanced BL21 E.coli derivative, chemically competent E. coli cells suitable for high efficiency transformation and for T7 expression with enhanced reduction of basal expression.