Gel Loading Dye Orange (6X), New England Biolabs
Supplier: New England Biolabs (NEB)
B7022S
101417-882EA 57 USD
101417-882
Gel Loading Dye Orange (6X), New England Biolabs
Stains Electrophoresis Stains
Gel Loading Dye, Orange (6X) is a pre-mixed loading buffer with a tracking dye for agarose and non-denaturing polyacrylamide gel electrophoresis
This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to their DNA substrates following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. Orange G will not appear in gel photographs, and runs ahead of all but the smallest restriction fragments. Orange G migrates at approximately 50 bp on a standard 1% TBE agarose gel.