Pipet-Aid™ Portable Pipet-Aid, Bellco Glass
Supplier: Bellco Glass
1225-85000
101416-104EA 763.55 USD
101416-104
Pipet-Aid™ Portable Pipet-Aid, Bellco Glass
Pipettes Pipette Controllers
Rechargeable self-contained unit that provides two hours of pipetting before recharging is necessary.
  • Each button has three speed settings (fast, medium, slow)
Delivery information: Includes battery charger, four replacement filters and tissue culture nose piece.