96-Well Collection Plates, Omega Bio-tek
Supplier: Omega Bio-Tek
EZ9602-02
101414-422CS 40.06 USD
101414-422 101414-424
96-Well Collection Plates, Omega Bio-tek
Microplates HTS Microplates
These well plates are manufactured from high polypropylene (PP) for excellent temperature and chemicals resistance.
  • Can be used as collection plate for E-Z 96 DNA/RNA plate
  • Fits to the vacuum manifold
  • Fits well with most robotic work stations