AeraSeal Film, Omega Bio-tek®
Supplier: Omega Bio-Tek
AC1201-02
101414-322PK 111.76 USD
101414-322
AeraSeal Film, Omega Bio-tek®
Seals Microplate Seals
A 114 µm thick, hydrophobic, porous film with medical-grade adhesive for tissue culture plates, bio-blocks and 96-well plates where gas exchange is necessary for cell or bacterial growth.
AeraSeal films minimize cross-contamination, spillage, and evaporation. They allow uniform air and CO2 exchange for all wells, unlike the plate lids which favor exchange for wells near plate edges.