dNTP Mix, 10 mM, QuantaBio
Supplier: Quantabio
95062-01K
101414-200EA 0 USD
101414-200 101414-198
dNTP Mix, 10 mM, QuantaBio
Nucleic Acid Reagents Nucleotides
The 10mM dNTP Mix is a solution of high purity deoxyribonucleoside triphosphates that has been functionally qualified for real-time quantitative PCR (qPCR)
  • High Purity
  • Functionally tested
  • Improves dynamic range, sensitivity and precision of qPCR assays
It is suitable for use in conventional end-point PCR, qPCR, first-strand cDNA synthesis, and other applications that require dNTP as substrate. Quality and purity of dNTP can directly affect dynamic range, sensitivity and precision of qPCR assays. The use of qPCR certified dNTPs is a simple but important measure to ensure consistent assay performance.