Large Dissecting Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72951-15
101413-594EA 56.96 USD
101413-594
Large Dissecting Kit, Electron Microscopy Sciences
Dissection Sets
This dissection kit includes the following:
  • 12 Scalpel Blades #11 with #3 Handle
  • 6 1/2" Kelly Hemostat, Straight
  • 6 ¼" Kelly Hemostat, Straight
  • 4 1/2" Iris Scissors
  • 51/2" Dissecting Scissors
  • Set of Dissecting Needle, one Straight and one Bent
  • 4 1/2" Straight Fine Point Serrated Jaws Forceps
  • 5 1/2" Straight Blunt tip Serrated Jaws Forceps
  • 2 mm Diameter Stainless Steel Punch
  • Suture Hook
  • Large Wood Handle Spatula EMS # Description Qty/Cs