Ear Punch, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72893-01
101413-586EA 46.61 USD
101413-586
Ear Punch, Electron Microscopy Sciences
Biopsy Punches
Made from stainless steel. Allows for you to make a 2 mm hole through the animal ear in just one second. It also can be used as a specimen puncher.