Wet Surface Pen, Black, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72128
101413-508EA 79.05 USD
101413-508
Wet Surface Pen, Black, Electron Microscopy Sciences
Pens Lab Markers
This unusual pen will write clearly on wet surfaces, such as marking slots on wet Southern, Northern, and Western blots Use it for marking alignment dots on blotting filters placed over agar plates containing bacterial colonies or phage plaques
Mark lanes on wet plots prior to cutting the filter into strips to be incubated separately with different antisera or probes. It can be used to correct or write on wet labels that were kept in a water bath, in liquid nitrogen or in a freezer at 0 degrees. The Wet Surface Pen is extremely useful for marking filters during processing under water-More Product Information