Quasar L.E.D. Lighting System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68107-70
101413-154EA 62.16 USD
101413-154
Quasar L.E.D. Lighting System, Electron Microscopy Sciences
Lights LED Lights, Lamps and Bulbs
QUASAR™ LS Kit includes lighting system frame, retro fit attachment pins, longer screw for OptiLOUPE™ fitted visors, 2 ʽAA’ batteries, and velcro strap.
  • L.E.D. Lighting System for Optivision Head Band Magnifiers
  • Custom fit lighting attachment for magnifying visions
  • Patent pending design utilizes the latest in LED technology
  • Reduces shadows and increases visibility in any environment
  • Lights up holes and deep cavities
  • Super long batteries life –light remains bright after 16 hours of straight use
  • Lasts up to 96 hours solid on a single set of batteries
  • Over 100,000 hour bulb life expectanct
QUASAR™ LS Kit includes lighting system frame, retro fit attachment pins, longer screw for OptiLOUPE™ fitted visors, 2 ʽAA’ batteries, and velcro strap.