Aldehyde/Silane Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63476-EB
101412-854PK 511.16 USD
101412-854 101412-856 101412-858
Aldehyde/Silane Slide, Electron Microscopy Sciences
Slides Microarray Slides
Offers a uniform coating of aldehydes for attaching amine-modified DNA.