Modular Blocks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63129-01 63129-07 63133-01 63129-06 63129-10 63131-05 63131-02 63132-50 63129-05 63133-02 63130-12
101412-736EA 128.63 USD
101412-736 101412-738 101412-748 101412-760 101412-764 101412-746 101412-750 101412-756 101412-754 101412-762 101412-744 101412-766 102094-502
Modular Blocks, Electron Microscopy Sciences
Block Heaters Blocks
Solid black anodized aluminum blocks for better heat retention.
Block includes thermometer well for monitoring block temperatures. Close contact of tubes to block walls, provides excellent heat transfer. Block puller included with all models. For use with all our brand modular dri-baths. Single block size: 3.75W×2.9D×2" H (9.5×7.4×5.1 cm).