1 Gal Paraffin Dispenser, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62839-20 62839-10
101412-656EA 6048.76 USD
101412-656 101412-654
1 Gal Paraffin Dispenser, Electron Microscopy Sciences
Paraffin Wax Trimmers and Dispensers
This dispenser provides immediate 'Wax on Tap' for use in pathology laboratories. Durable construction of an anodized black interior stove, enameled black and white aluminum exterior and a black metal lid.
  • Thermally insulated tap heater with independent temperature contol eliminates wax solidification.
  • Built in controller provides temperature control from ambient to 75 °C
  • A non-drip swivel lever with replaceable filter makes the dispensing of wax easy
  • Safety over-temperature at 105 °C; non-resettable thermal fuse
  • Easy to clean
Capacity of 1 gal. (4.5 liters)