Bacti-Cinerator IV, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62420-10 62420-55 62420-54 62420-20 62420-51
101412-622EA 1100.38 USD
101412-622 101412-626 101412-636 101412-634 101412-624 101412-628
Bacti-Cinerator IV, Electron Microscopy Sciences
Sterilizers Loop Sterilizers
The Baci-Cinerator IV sterilizes by infrared heat (Heater Element contains no asbestos) - organic material is incinerated deep inside the ceramic tunnel. To use, simply insert loop or needle into the tunnel. Complete sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500 °F.
  • Prevents infectious spatter and/or cross contamination
  • No open flame -no hazardous gas
  • No need to wait for the loops or needles to turn red
  • Does not require oxygen so it can be used in anaerobic chambers
  • UL approved