Boron carbide, powder 42 µm

Supplier: Electron Microscopy Sciences
Warning

50510-42
101411-948EA 97.14 USD
101411-948
Boron carbide, powder 42 µm
Boron carbide

Boron Carbide is one of the hardest materials known, ranking third behind diamond and cubic boron nitride.


  • Extreme hardness, (Knoop 100g: 2900-3580 kg/mm2)
  • Good chemical resistance
  • Good nuclear properties
  • Low density (2.52 g/cm2)


Due to the hardness, boron carbide powder is used as an abrasive in polishing and lapping applications.

Formula: CB₄
MW: 55.25
Melting Pt: 2350
Density: 2.52
MDL Number: MFCD00011520
CAS Number: 12069-32-8
Merck Index: 12,01374

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR