Eosin Y 1% alcoholic solution stain for histology

Supplier: Electron Microscopy Sciences
Danger

26762-01
101411-522EA 74.84 USD
101411-522
Eosin Y 1% alcoholic solution stain for histology
Eosin Y
Formula: C₂₀H₈Br₄O₅
MDL Number: MFCD00036189
CAS Number: 15086-94-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76e6c20378da02247ac0eff1b31a1328f47ccd84&vid=8107c571244a0f3998d27678ac646c7199878ce2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR