Sudan IV in isopropanol (IPA) fat and lipid stain

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol

26503-01
101411-410EA 177.79 USD
101411-410
Sudan IV in isopropanol (IPA) fat and lipid stain
Sudan IV

For fat and lipid stain.


Stain results for Fat: Deep orange-red with Oil Red O, lighter with Sudan IV, Orange-yellow with Sudan II

Formula: C₂₄H₂₀N₄O
MW: 380.45 g/mol
Flash Pt: 13 °C (55.4 °F)
MDL Number: MFCD00003893
CAS Number: 85-83-6
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR