Wright-Giemsa stain
Supplier: Electron Microscopy Sciences
Synonyms:
Wright-Giemsa Solution
26161-02
101411-136EA
78.33
USD
101411-136
Wright-Giemsa stain
Wright-Giemsa stain (solutions & kits)
Used in Prussian Blue Stain for Non-Hemoglobin Iron Sundberg and Broman (1955). Stain results - Particulate Iron and Non-granular cytoplasm of many Macrophages: Blue to Blue-green Other: Comparable to original films stained.
Clark, G.: Staining Procedures, Williams & Wilkins Co. Baltimore, 3rd Ed., c.1973, p.130. Sunberg,. R.D. and Broman, H., Blood, 10160-6, 1955.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)