Wright-Giemsa stain for hematology

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Wright-Giemsa Solution

26149-01
101411-114EA 106.93 USD
101411-114
Wright-Giemsa stain for hematology
Wright-Giemsa stain (solutions & kits)

Wright-Giemsa Stain, Lillie (1972) is used for differentiation of blood corpuscles


Stain results
Erthrocytes: Pink-Tan
Eosinophiles-Granules: Red
Lymphocytes-Granules: Red-Purple
Cytoplasm: Blue
Neutrophils-Granules: Purple-Pink


To use with phosphate buffer pH 6.5 (26149-02)

MDL Number: MFCD00132593
UN: 1230
ADR: 3 (6.1) II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR