Acetone:Alcohol (1:1) for Gram staining
Supplier: Electron Microscopy Sciences
Danger
26105-5A
101411-044EA 27.53 USD
101411-044
Acetone:Alcohol (1:1) for Gram staining
Acetone:Alcohol
Reagent of Brown and Brenn Method for Gram Positive and Gram Negative Bacteria
Gram Negative Bacteria for bacteria stain: Gram + / Gram – bacteria and filaments of nocardia, etc
Stain results - Gram + Bacteria, Nocardia and Actinomyces Filaments: Blue, Gram – Bacteria, Nuclei: Red, Additional tissue elements: Yellow
UN: 1993
ADR: 3,II