Hematoxylin 10% in alcohol

Supplier: Electron Microscopy Sciences

26091-10
101411-012EA 187.51 USD
101411-012
Hematoxylin 10% in alcohol
Hematoxylin

Alcoholic stain solutions are available with Congo Red 0.2%, Hematoxylin 1% in 95% alcohol, Iodine 0.5% in 95% alcohol, Iodine 0.5% in 85% alcohol, Iodine 2% in 85% alcohol, Saffron 6%, Thionin 0.5% in 20% alcohol, Aldehyde Fuchsin and Alum Hematoxylin.

Formula: C₁₆H₁₄O₆
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR