Hematoxylin 1% in ethanol 95%

Supplier: Electron Microscopy Sciences

Synonyms: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin

26091-01
101411-010EA 65.65 USD
101411-010
Hematoxylin 1% in ethanol 95%
Hematoxylin

Alcoholic stain solutions are available with Congo Red 0.2%, Hematoxylin 1% in 95% alcohol, Iodine 0.5% in 95% alcohol, Iodine 0.5% in 85% alcohol, Iodine 2% in 85% alcohol, Saffron 6%, Thionin 0.5% in 20% alcohol, Aldehyde Fuchsin and Alum Hematoxylin.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR