Congo red 0.5% in aqueous solution

Supplier: Electron Microscopy Sciences
Danger

26090-20
101411-006EA 48 USD
101411-006
Congo red 0.5% in aqueous solution
Congo Red

  • Premixed ready-to-use; saves time and effort
  • Consistently uniform results are obtained
  • Packaged and prepared in convenient and precise quantities

Formula: C₃₂H₂₂N₆Na₂O₆S₂
MDL Number: MFCD00004028
CAS Number: 573-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR