Cresyl violet (acetate) 2% in aqueous solution stain for histology

Supplier: Electron Microscopy Sciences
Warning

26089-20
101411-002EA 178.88 USD
101411-002
Cresyl violet (acetate) 2% in aqueous solution stain for histology
Cresyl violet (acetate)

Cresyl Violet Acetate solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology.


Available in concentrations of 0.1% aqueous or 2.0% aqueous.


Certifications: Certificates of analysis are available.

Formula: C₁₈H₁₅N₃O₃
MDL Number: MFCD00013151
CAS Number: 10510-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e3a3e2b7912b5c3357b004bb83294a1587f06a5&vid=b3c2c537ac0ed39530077aa0a3a99201a9ec4217&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR