Crystal violet 1% Stirling's solution

Supplier: Electron Microscopy Sciences

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

26088-18
101410-998EA 62.18 USD
101410-998
Crystal violet 1% Stirling's solution
Crystal Violet

Crystal Violet solutions are prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology


Available in a concentrations of 1.0% aqueous or Huckers solution, Lieb's stock solution, or Stirling's solution.

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
Merck Index: 13,04406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c35cef669eed8b365e5e7cebd131e956492ec0bb&vid=594b87fb4bb6be75ea109318c578f12c35c37bad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR