Leishman's eosin-methylene blue

Supplier: Electron Microscopy Sciences

Synonyms: Eosin and methylene blue Leishman, Eosin-Methylene blue dye Leishman, Leishman's stain, Leishman stain, Leishman's stain (eosin methylene blue compound)

26086-05
101410-986EA 121.44 USD
101410-986
Leishman's eosin-methylene blue
Leishman's Eosin-Methylene blue

Lieshman’s Stain and Working Buffer

MDL Number: MFCD00131498
CAS Number: 12627-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR