Methylene blue working solution

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

26075-20
101410-964EA 53.37 USD
101410-964
Methylene blue working solution
Methylene blue

  • High quality staining solution
  • Ready-to-use
  • Working solution, aqueous or in alchohol


Methylene Blue solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology. Available in concentrations of 1.0% aqueous, 1.4% in alcohol and as working solution.

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
MDL Number: MFCD00150008
CAS Number: 61-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR