Toluidine blue O 0,1% in aqueous solution biological stain

Supplier: Electron Microscopy Sciences

Synonyms: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

26074-05
101410-954EA 46.75 USD
101410-954
Toluidine blue O 0,1% in aqueous solution biological stain
Toluidine blue O

Toluidine Blue O Aqueous solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology


Available in concentrations 0.1% and 1% aqueous solution.

Formula: C₁₅H₁₆ClN₃S
MDL Number: MFCD00011934
CAS Number: 92-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98cdbd73b0eae5397b34d0b674293097b26557db&vid=14ea2f9a0c3a3d18b8feddbe1a7931c160340c03&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR