Safranine O 0.1% in 1% acetic acid

Supplier: Electron Microscopy Sciences
Warning

26069-05
101410-950EA 40.7 USD
101410-950
Safranine O 0.1% in 1% acetic acid
Safranine O

Safranin O Solution in Acetic Acid is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology


Available in concentrations 0.1% in 0.1% acetic acid and 0.2% in 1% Acetic Acid.

Formula: C₂₀H₁₉ClN₄
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR