Orange G 0,5% in aqueous solution

Supplier: Electron Microscopy Sciences

26066-05
101410-940EA 112.92 USD
101410-940
Orange G 0,5% in aqueous solution
Orange G
Formula: C₁₆H₁₀N₂Na₂O₇S₂
MDL Number: MFCD00012457
CAS Number: 1936-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75239c050046ddf2bf82a771aa686f72ccb9dcac&vid=f1fc865635f1d8dfe800adcf738a98714cf8098d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR