Solvent Blue 38 0.1% in 95% alcohol

Supplier: Electron Microscopy Sciences

Synonyms: Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN

26056-15
101410-936EA 65.65 USD
101410-936
Solvent Blue 38 0.1% in 95% alcohol
Solvent Blue 38

Luxol Fast Blue solution Electron Microscopy Sciences is available in a concentration of 0.1% in 95% alcohol with acetic acid and a concentration of 0.1% in 95% alcohol.

Formula: C32H12CuN8Na2O6S2
MW: 778.15 g/mol
MDL Number: MFCD00071424
CAS Number: 1328-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32316b570c11ee4bd0ddfb0c40af5bcb9fd43d07&vid=c83bc63f3f9b4ce96069988bfd596f9be5ece55d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR