Hematoxylin 1% in aqueous solution

Supplier: Electron Microscopy Sciences

26042-1
101410-910EA 62.69 USD
101410-910
Hematoxylin 1% in aqueous solution
Hematoxylin

Hematoxylin solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the biological staining commission and the armed forces institute of pathology.


  • Available in concentrations of 1.0% aqueous, 5% in SD alcohol, in 95% alcohol, and as Heidenhain's hematoxylin

Formula: C₁₆H₁₄O₆
Boiling Pt: 100 °C (212 °F)
MDL Number: MFCD00078111
CAS Number: 517-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41846ee8d4e9428f4f96bbb3267844dd6778c6a7&vid=199dc970eeaf2ff9995b541a44efb8d8569ffe82&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR