Basic fuchsin 0.1% in methanol 95%

Supplier: Electron Microscopy Sciences

Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

26031-10
101410-892EA 89.45 USD
101410-892
Basic fuchsin 0.1% in methanol 95%
Basic Fuchsin

Basic Fuchsin solution Electron Microscopy Sciences is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology.
Available in concentrations of 0.25% aqueous, 0.5% aqueous, 0.1% in 95% methanol, and as a working solution.

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
MDL Number: MFCD00012569
CAS Number: 632-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6286f499dcea0ba2be8463e133aa74a5a9b8aa4&vid=11d895ce8f97553fbb42b1edca4d84c9b282f9ae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR