Alcian blue 1% in 1% acetic acid

Supplier: Electron Microscopy Sciences

26026-11
101410-876EA 128.16 USD
101410-876
Alcian blue 1% in 1% acetic acid
Alcian blue 8 GX

Alcian Blue Solution, Electron Microscopy Sciences, is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology. Available in concentrations of 1.0% aqueous, and 1.0% aqueous in 1% or 3% acetic acid.

Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number: MFCD00010720
CAS Number: 33864-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR