Acridine orange (hemizinc salt) 0.2% in aqueous solution

Supplier: Electron Microscopy Sciences

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

26024-02
101410-872EA 79.33 USD
101410-872
Acridine orange (hemizinc salt) 0.2% in aqueous solution
Acridine orange (hemizinc salt)

Electron Microscopy Sciences Acridine Orange Aqueous solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology. Available in concentrations of 0.1% aqueous and 0.2% aqueous.

Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MW: 369.96 g/mol
MDL Number: MFCD00081043
CAS Number: 10127-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR