Potassium hexacyanoferrate(III) 20% in aqueous solution

Supplier: Electron Microscopy Sciences

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

25102-20
101410-848EA 72.35 USD
101410-848
Potassium hexacyanoferrate(III) 20% in aqueous solution
Potassium hexacyanoferrate (III)

Prepared with ACS reagent

Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2

Order Now

Specification Test Results

Chloride 0.01%
Ferro compounds (Fe(Cn)₄ 0.05%
Insoluble matters 0.005%
Sulfate 0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR