Sodium hydroxide 3,1% in aqueous solution
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
21172-3
101410-804EA
55.85
USD
101410-804
Sodium hydroxide 3,1% in aqueous solution
Sodium hydroxide
- Sodium hydroxide solution is available in concentrations of 1.0%, 2.5%, 3.1% and 10.0% aqueous
Formula:
NaOH |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...