Potassium permanganate 0.3% in aqueous solution

Supplier: Electron Microscopy Sciences

20203
101410-778EA 45.65 USD
101410-778
Potassium permanganate 0.3% in aqueous solution
Potassium permanganate
Formula: KMnO₄
MDL Number: MFCD00011364
CAS Number: 7722-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR