Potassium dichromate 3% in aqueous solution

Supplier: Electron Microscopy Sciences

20103-3
101410-766EA 57.58 USD
101410-766
Potassium dichromate 3% in aqueous solution
Potassium dichromate

Potassium Dichromate Aqueous Electron Microscopy Sciences solutions are available in concentrations of 3.0% aqueous and 5.0% aqueous.

Formula: K₂Cr₂O₇
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 3287
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR