Hydroquinone 1% in aqueous solution

Supplier: Electron Microscopy Sciences

16795-1
101410-696EA 101.46 USD
101410-696
Hydroquinone 1% in aqueous solution
Hydroquinone

Hydroquinone Aqueous solution is available in concentrations of 1% aqueous, 2% aqueous, 4.0% aqueous.

Formula: C₆H₄(OH)₂
MDL Number: MFCD00002339
CAS Number: 123-31-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5821135d4ce76cb024d02a740e485f53ef702d32&vid=7f898d99dde47ccd203c7cd1f4dd3fb0b96c1c9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR