Hexamethylenetetramine 3% in aqueous solution

Supplier: Electron Microscopy Sciences

16712
101410-684EA 44.69 USD
101410-684
Hexamethylenetetramine 3% in aqueous solution
Methenamine
Formula: C₆H₁₂N₄
MDL Number: MFCD00006895
CAS Number: 100-97-0
UN: 1328
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3d096d3ddc6c104be37a7b4416853824225ea6a&vid=5b5d52a9970fcc32e47a733d1c73edcaef4213a2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR