Iron(III) chloride 2% in aqueous solution for histology

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Iron trichloride, Ferric chloride

15512-02
101410-656EA 65.81 USD
101410-656
Iron(III) chloride 2% in aqueous solution for histology
Iron (III) chloride
Formula: FeCl₃
MDL Number: MFCD00011005
CAS Number: 7705-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c542182b5d09fa972a328e993af03cf76c28cc7f&vid=4220e4a6411303112e0c7587cdb8a6ac63b9e57c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR