Acetone fixative reagent, buffered

Supplier: Electron Microscopy Sciences

10018
101410-606EA 55.12 USD
101410-606
Acetone fixative reagent, buffered
Acetone

Acetone

Formula: CH₃COCH₃
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR