Breathe-Easy® Sealing Film for Petri Dishes, Diversified Biotech
Supplier: Diversified Biotech
BEMP-1000
10141-850PK 123.78 USD
10141-850
Breathe-Easy® Sealing Film for Petri Dishes, Diversified Biotech
Seals Microplate Seals
Strip design allows film to be protected during application; a tab remains attached for easy removal.
  • Strips allow for beneficial exchange of oxygen* and CO2 for cellular growth
  • Prevent microbial and viral contamination during growth and storage of cells
  • Dishes sealed with Breathe Easy strips may be incubated and/or stored at ambient temperatures, refrigerated or frozen