Breathe-Easier Sealing Film, Diversified Biotech
Supplier: Diversified Biotech
BERM-2000
10141-844PK 101.88 USD
10141-844
Breathe-Easier Sealing Film, Diversified Biotech
Seals Microplate Seals
Allows rapid and uniform transmission of oxygen, carbon dioxide and water vapors to stimulate cell growth.
  • Non-cytotoxic and sterile - Membranes are free of cytotoxins and also gamma irradiated to assure sterility
  • Chemically and thermally stable - Resists a majority of caustic agents and organic solvents as well as exposure to temperatures from -20°C to 80°C
  • Easy sample retrieval - membrane is easily pierced by pipet tips
  • US Patent # 5,858,770