Anti-CD86 Rat Antibody [clone: GL-1 (GL1)]

Supplier: Tonbo Biosciences
40-0862-U100
10141-164EA 82.5 USD
10141-164
Anti-CD86 Rat Antibody [clone: GL-1 (GL1)]
Antibodies
The GL-1 antibody reacts with mouse CD86, also known as B7-2, an 80 kDa cell surface protein which is a ligand for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 can also bind a second B7 ligand known as CD80 (B7-1). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response. 

The GL-1 antibody may be used as a marker for CD86 expression on B cells, macrophages, and dendritic cells.

Type:
Antigen: CD86
Clonality:
Clone: GL-1 (GL1)
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR