Anti-CD18 Rat Monoclonal Antibody [clone: M18/2]

Supplier: Biolegend
101401 101402
101401-BLEA 60.8 USD
101401-BL 101402-BL
Anti-CD18 Rat Monoclonal Antibody [clone: M18/2]
Antibodies
Purified anti-mouse CD18 [M18/2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: M18/2
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR