MicroMist™ Nebulizer for ELAN/NexION, PerkinElmer
Supplier: PerkinElmer
N0775342
10140-322EA 2934.47 USD
10140-322
MicroMist™ Nebulizer for ELAN/NexION, PerkinElmer
Atomizers
This glass MicroMist™ nebulizer includes connectors and sample tubing for use with ELAN® 5000/6x00/9000/DRCs and NexION® 300 and 350 ICP-MS series instruments. It can also be used with Optima® 3x00 RL ICP-OES series instruments.
The MicroMist™ micro-uptake glass concentric nebulizer offers high performance for limited sample volumes. MicroMist™ for ELAN/NexION 300/350 are fitted with a UniFit connector with 1.3×0.50×700 (I.Ø×O.Ø×L) mm sample tubing.