SeaSpray™ Glass Nebulizer for ELAN® and NexION® 300/350, PerkinElmer
Supplier: PerkinElmer
N0775340
10140-318EA 3166 USD
10140-318
SeaSpray™ Glass Nebulizer for ELAN® and NexION® 300/350, PerkinElmer
Atomizers
This glass SeaSpray™ nebulizer is used with ELAN® 5000/6x00/9000/DRCs and NexION® 300 and 350 ICP-MS series instruments.
  • Glass construction
  • 2.0 ml/min. uptake rate
  • Supplied with the EzyLok argon connector
The SeaSpray™ concentric nebulizer is ideal for samples with high dissolved solids, and offers outstanding nebulization efficiency for trace level analysis. It offers freedom from clogging while nebulizing solutions near the limit of solubility of most mineral salts, and conferring significant sensitivity gains. They are supplied with the EzyLok argon connector.